1-(2-(Methoxymethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

35747-1G 35747-25G 35747-500MG 35747-5G
76888-076EA 333.13 USD
76888-076 76888-078 76888-080 76888-082
1-(2-(Methoxymethyl)phenyl)ethanone ≥97%
1-(2-(Methoxymethyl)phenyl)ethanone
Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24690358
CAS Number: 24807-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR