(4-Fluoronaphthalen-1-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

35551-1G 35551-25G 35551-5G
76888-056EA 80.63 USD
76888-056 76888-058 76888-060
(4-Fluoronaphthalen-1-yl)methanol ≥97% research grade
(4-Fluoronaphthalen-1-yl)methanol
Formula: C₁₁H₉FO
MW: 176.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059454
CAS Number: 79996-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR