2,4-Dichloro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35290-1G 35290-25G 35290-5G 35290-500MG
76888-014EA 100.79 USD
76888-014 76888-016 76888-018 77650-308
2,4-Dichloro-3-hydroxybenzaldehyde ≥97%
2,4-Dichloro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.011 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22199563
CAS Number: 56962-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR