2-(Benzyloxy)-1-bromo-4-methylbenzene 95%

Supplier: AOBChem USA

Warning

35270-1G 35270-25G 35270-5G 35270-500MG
76888-002EA 117.76 USD
76888-002 76888-004 76888-006 77650-298
2-(Benzyloxy)-1-bromo-4-methylbenzene 95%
2-(Benzyloxy)-1-bromo-4-methylbenzene
Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28014039
CAS Number: 177555-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR