1-(Benzyloxy)-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

35091-1G 35091-25G 35091-5G
76887-984EA 76.39 USD
76887-984 76887-986 76887-988
1-(Benzyloxy)-2-fluorobenzene ≥97%
1-(Benzyloxy)-2-fluorobenzene
Formula: C₁₃H₁₁FO
Storage Temperature: Refrigerator
MDL Number: MFCD09032646
CAS Number: 368-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR