1-Bromo-2-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

34967-1G 34967-25G 34967-5G
76887-972EA 62.6 USD
76887-972 76887-974 76887-976
1-Bromo-2-fluoro-3-iodobenzene ≥97%
1-Bromo-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
Storage Temperature: Refrigerator
MDL Number: MFCD13194483
CAS Number: 958458-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR