2,3,5-Trichloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Iodo-2,3,5-trichloropyridine

34899-1G 34899-25G 34899-500MG 34899-5G
76887-958EA 126.25 USD
76887-958 76887-960 76887-962 76887-964
2,3,5-Trichloro-4-iodopyridine ≥97%
2,3,5-Trichloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅HCI₃IN
Storage Temperature: Refrigerator
CAS Number: 406676-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR