1,4-Dibromo-2-chloro-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

34813-25G 34813-5G
76887-932EA 524.09 USD
76887-932 76887-934
1,4-Dibromo-2-chloro-3-fluorobenzene ≥95%
1,4-Dibromo-2-chloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
Storage Temperature: Ambient
MDL Number: MFCD09878168
CAS Number: 1000573-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR