(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

34791-1G 34791-25G 34791-500MG 34791-5G
76887-916EA 352.22 USD
76887-916 76887-918 76887-920 76887-922
(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid ≥95%
(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BFNO₂
MW: 197.014 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27935575
CAS Number: 1704063-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR