(3-(Pyridin-3-yl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-(3-Pyridyl)benzeneboronic acid, [3-(Pyridin-3-yl)phenyl]boronic acid

34726-25G 34726-5G
76887-906EA 595.17 USD
76887-906 76887-908
(3-(Pyridin-3-yl)phenyl)boronic acid ≥97% research grade
(3-(Pyridin-3-yl)phenyl)boronic acid
Formula: C₁₁H₁₀BNO₂
MW: 199.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658918
CAS Number: 351422-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR