2,4,6-Bribromobenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

34650-1G 34650-25G 34650-500MG 34650-5G
76887-890EA 155.96 USD
76887-890 76887-892 76887-894 76887-896
2,4,6-Bribromobenzoic acid methyl ester ≥97%
2,4,6-Bribromobenzoic acid methyl ester
Formula: C₈H₅Br₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18399688
CAS Number: 14920-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR