2,3-Difluoro-4-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34607-1G 34607-25G 34607-5G
76887-878EA 62.6 USD
76887-878 76887-880 76887-882
2,3-Difluoro-4-hydroxybenzoic acid ≥97%
2,3-Difluoro-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01321116
CAS Number: 175968-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR