(3-Methoxy-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

34395-25G 34395-5G
76887-856EA 275.84 USD
76887-856 76887-858
(3-Methoxy-4-methylphenyl)methanol ≥97%
(3-Methoxy-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797828
CAS Number: 4685-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR