6-Chloro-2,3-difluorotoluene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Difluoro-6-chlorotoluene

34335-1G 34335-25G 34335-500MG 34335-5G
76887-830EA 285.39 USD
76887-830 76887-832 76887-834 76887-836
6-Chloro-2,3-difluorotoluene ≥95%
6-Chloro-2,3-difluorotoluene
Formula: C₇H₅ClF₂
MW: 162.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922582
CAS Number: 1208077-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR