1,2-Dichloro-3-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

34333-1G 34333-25G 34333-500MG 34333-5G
76887-818EA 260.99 USD
76887-818 76887-820 76887-822 76887-824
1,2-Dichloro-3-fluoro-4-iodobenzene ≥97%
1,2-Dichloro-3-fluoro-4-iodobenzene
Formula: C₆H₂Cl₂FI
MW: 290.889 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13194305
CAS Number: 1188535-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR