2,4-Dichloro-3-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

34235-1G 34235-25G 34235-500MG 34235-5G
76887-798EA 379.81 USD
76887-798 76887-800 76887-802 76887-804
2,4-Dichloro-3-fluorophenol ≥97%
2,4-Dichloro-3-fluorophenol
Formula: C₆H₃Cl₂FO
MW: 180.992 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24722775
CAS Number: 343-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR