2,3-Dichlorothioanisole 95%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dichloro-3-(methylsulphanyl)benzene, 2,3-Dichlorophenyl methyl sulphide, 1,2-Dichloro-3-(methylthio)benzene

34123-1G 34123-25G 34123-5G 34123-500MG
76887-774EA 81.69 USD
76887-774 76887-776 76887-778 77646-220
2,3-Dichlorothioanisole 95%
2,3-Dichlorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆Cl₂S
Storage Temperature: Refrigerator
MDL Number: MFCD03789169
CAS Number: 17733-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR