1-Bromo-2,3-dichloro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

34077-1G 34077-25G 34077-500MG 34077-5G
76887-750EA 304.48 USD
76887-750 76887-752 76887-754 76887-756
1-Bromo-2,3-dichloro-4-methoxybenzene ≥97%
1-Bromo-2,3-dichloro-4-methoxybenzene
Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
Storage Temperature: Ambient
CAS Number: 109803-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR