(2,5-Difluoro-4-phenylmethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

34014-1G 34014-500MG 34014-5G
76887-716EA 524.09 USD
76887-716 76887-718 76887-720
(2,5-Difluoro-4-phenylmethoxyphenyl)methanol ≥97%
(2,5-Difluoro-4-phenylmethoxyphenyl)methanol
Formula: C₁₄H₁₂F₂O₂
MW: 250.241 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28503328
CAS Number: 1936712-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR