(S)-2,2'-Dihydroxy-1,1'-binaphthalene-3,3'-diboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

33852-1G 33852-500MG 33852-5G
76887-702EA 589.87 USD
76887-702 76887-704 76887-706
(S)-2,2'-Dihydroxy-1,1'-binaphthalene-3,3'-diboronic acid pinacol ester ≥97% research grade
(S)-2,2'-Dihydroxy-1,1'-binaphthalene-3,3'-diboronic acid pinacol ester
Formula: C₃₂H₃₆B₂O₆
Storage Temperature: Ambient
MDL Number: MFCD31561341
CAS Number: 1638840-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR