1-Bromo-2-chloro-4,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

33656-1G 33656-25G 33656-500MG 33656-5G
76887-648EA 206.88 USD
76887-648 76887-650 76887-652 76887-654
1-Bromo-2-chloro-4,5-dimethylbenzene ≥97%
1-Bromo-2-chloro-4,5-dimethylbenzene
Formula: C₈H₈BrCl
MW: 219.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922557
CAS Number: 500536-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR