2-((4-Methylpiperazin-1-yl)methyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33621-1G 33621-25G 33621-500MG 33621-5G
76887-636EA 173.99 USD
76887-636 76887-638 76887-640 76887-642
2-((4-Methylpiperazin-1-yl)methyl)phenylboronic acid ≥97%
2-((4-Methylpiperazin-1-yl)methyl)phenylboronic acid
Formula: C₁₂H₁₉BN₂O₂
MW: 234.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16198079
CAS Number: 1312921-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR