1,3-Dibromo-5-butoxybenzene ≥95%

Supplier: AOBChem USA

Warning

33603-1G 33603-25G 33603-500MG 33603-5G
76887-624EA 260.99 USD
76887-624 76887-626 76887-628 76887-630
1,3-Dibromo-5-butoxybenzene ≥95%
1,3-Dibromo-5-butoxybenzene
Formula: C₁₀H₁₂Br₂O
Storage Temperature: Refrigerator
MDL Number: MFCD29043831
CAS Number: 896745-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR