1-Bromo-4,5-difluoro-2-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

33480-25G 33480-5G
76887-612EA 545.31 USD
76887-612 76887-614
1-Bromo-4,5-difluoro-2-(phenylmethoxy)benzene ≥97%
1-Bromo-4,5-difluoro-2-(phenylmethoxy)benzene
Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619446
CAS Number: 1698908-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR