2-(4-Bromophenyl)-1,1,1-trifluoropropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

33086-1G 33086-25G 33086-500MG 33086-5G
76887-534EA 215.37 USD
76887-534 76887-536 76887-538 76887-540
2-(4-Bromophenyl)-1,1,1-trifluoropropan-2-ol ≥97%
2-(4-Bromophenyl)-1,1,1-trifluoropropan-2-ol
Formula: C₉H₈BrF₃O
MW: 269.058 g/mol
Storage Temperature: Refrigerator
CAS Number: 122243-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR