2,3-Dibromobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32855-25G 32855-5G
76887-494EA 488.02 USD
76887-494 76887-496
2,3-Dibromobenzoic acid ≥97%
2,3-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
MW: 279.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520726
CAS Number: 603-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR