(4-Chloro-2,5-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

32790-1G 32790-25G 32790-500MG 32790-5G
76887-480EA 350.1 USD
76887-480 76887-482 76887-484 76887-486
(4-Chloro-2,5-difluorophenyl)methanol ≥97%
(4-Chloro-2,5-difluorophenyl)methanol
Formula: C₇H₅ClF₂O
MW: 178.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847043
CAS Number: 252004-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR