2,4-Dichloro-6-fluorophenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

32686-1G 32686-25G 32686-500MG 32686-5G
76887-464EA 379.81 USD
76887-464 76887-466 76887-468 76887-470
2,4-Dichloro-6-fluorophenylboronic acid pinacol ester ≥97% research grade
2,4-Dichloro-6-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₄BCl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12405366
CAS Number: 1192002-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR