(4-Formylthiophen-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

32392-1G 32392-500MG 32392-5G 32392-25G
76887-438EA 495.45 USD
76887-438 76887-440 76887-442 77655-042
(4-Formylthiophen-2-yl)boronic acid ≥97%
(4-Formylthiophen-2-yl)boronic acid
Formula: C₅H₅BO₃S
MW: 155.967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383061
CAS Number: 1186026-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR