2-Benzyloxy-1-chloro-3-fluorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-1-chloro-3-fluorobenzene

32155-25G 32155-5G
76887-412EA 453.01 USD
76887-412 76887-414
2-Benzyloxy-1-chloro-3-fluorobenzene ≥97% research grade
2-Benzyloxy-1-chloro-3-fluorobenzene
Formula: C₁₃H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD13195733
CAS Number: 938180-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR