2-(Benzyloxy)isonicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

32015-1G 32015-25G 32015-500MG 32015-5G
76887-380EA 215.37 USD
76887-380 76887-382 76887-384 76887-386
2-(Benzyloxy)isonicotinic acid ≥97% research grade
2-(Benzyloxy)isonicotinic acid
Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100038
CAS Number: 467236-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR