2-(2-Brom-3-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

31990-25G 31990-5G
76887-362EA 364.95 USD
76887-362 76887-364
2-(2-Brom-3-fluorophenyl)-1,3-dioxolane ≥97%
2-(2-Brom-3-fluorophenyl)-1,3-dioxolane
Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619435
CAS Number: 2169687-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR