2,3-Dimethoxy-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethoxypyridine-5-boronic acid, pinacol ester, 2,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

31898-1G 31898-25G 31898-500MG 31898-5G
76887-346EA 221.73 USD
76887-346 76887-348 76887-350 76887-352
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester ≥97%
2,3-Dimethoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD11857651
CAS Number: 1083168-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR