2-(Difluoromethoxy)-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31838-1G 31838-25G 31838-500MG 31838-5G
76887-330EA 287.51 USD
76887-330 76887-332 76887-334 76887-336
2-(Difluoromethoxy)-5-fluorobenzaldehyde ≥97%
2-(Difluoromethoxy)-5-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698502
CAS Number: 1214326-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR