1-Bromo-2,4-difluoro-5-methoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
31724-1G
31724-25G
31724-500MG
31724-5G
76887-316EA
272.66
USD
76887-316
76887-318
76887-320
76887-322
1-Bromo-2,4-difluoro-5-methoxybenzene ≥97%
1-Bromo-2,4-difluoro-5-methoxybenzene
Formula:
C₇H₅BrF₂O MW: 223.0148 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28401114 CAS Number: 943831-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...