1-(3,5-Dichloropyridin-4-yl)ethan-1-one ≥97% research grade

Supplier: AOBChem USA

Warning

31603-1G 31603-500MG 31603-5G 31603-25G
76887-302EA 510.3 USD
76887-302 76887-304 76887-306 77654-946
1-(3,5-Dichloropyridin-4-yl)ethan-1-one ≥97% research grade
1-(3,5-Dichloropyridin-4-yl)ethan-1-one
Formula: C₇H₅Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD11847308
CAS Number: 402561-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR