4-Bromo-2,5-difluorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,5-difluoro-4-methylbenzene

31505-1G 31505-25G 31505-5G
76887-290EA 94.43 USD
76887-290 76887-292 76887-294
4-Bromo-2,5-difluorotoluene ≥97%
4-Bromo-2,5-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368736
CAS Number: 252004-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR