1-Chloro-4,5-difluoro-2-methoxybenzene ≥95% research grade

Supplier: AOBChem USA

Warning

31477-1G 31477-25G 31477-500MG 31477-5G
76887-278EA 328.88 USD
76887-278 76887-280 76887-282 76887-284
1-Chloro-4,5-difluoro-2-methoxybenzene ≥95% research grade
1-Chloro-4,5-difluoro-2-methoxybenzene
Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD18415542
CAS Number: 1261598-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR