2,3-Dihydro-1,4-benzodioxin-6-ylmethanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dihydro-6-(hydroxymethyl)-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanol

31430-25G 31430-5G
76887-266EA 418 USD
76887-266 76887-268
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol ≥97%
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317581
CAS Number: 39270-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR