2-Chloro-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-3-methylphenyl)boronic acid

31378-1G 31378-25G 31378-5G 31378-500MG
76887-254EA 96.55 USD
76887-254 76887-256 76887-258 77649-994
2-Chloro-3-methylphenylboronic acid ≥97%
2-Chloro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₂
MW: 170.4000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797215
CAS Number: 915070-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR