1-Bromo-3-chloro-2,4-dimethylbenzene 95%

Supplier: AOBChem USA

Warning

31335-1G 31335-25G 31335-5G 31335-500MG
76887-244EA 76.39 USD
76887-244 76887-246 76887-248 77645-968
1-Bromo-3-chloro-2,4-dimethylbenzene 95%
1-Bromo-3-chloro-2,4-dimethylbenzene
Formula: C₈H₈BrCl
MW: 219.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079711
CAS Number: 1782344-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR