[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)methanol

31283-25G 31283-5G
76887-232EA 272.66 USD
76887-232 76887-234
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol ≥97%
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol
Formula: C₁₄H₁₁F₃O
MW: 252.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862515
CAS Number: 457889-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR