2,3-Difluoro-4-(phenylmethoxy)phenol ≥97%
Supplier: AOBChem USA
|
Warning
|
31274-1G
31274-25G
31274-500MG
31274-5G
76887-220EA
260.99
USD
76887-220
76887-222
76887-224
76887-226
2,3-Difluoro-4-(phenylmethoxy)phenol ≥97%
2,3-Difluoro-4-(phenylmethoxy)phenol
Formula:
C₁₃H₁₀F₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD23110933 CAS Number: 271254-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...