1-Bromo-4-chloro-3-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

31257-1G 31257-25G 31257-500MG 31257-5G
76887-192EA 292.81 USD
76887-192 76887-194 76887-196 76887-198
1-Bromo-4-chloro-3-fluoro-2-methylbenzene ≥95%
1-Bromo-4-chloro-3-fluoro-2-methylbenzene
Formula: C₇H₅BrClF
Storage Temperature: Refrigerator
MDL Number: MFCD28141988
CAS Number: 1628524-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR