2,4-Dichloro-3-fluorobromobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,4-dichloro-3-fluorobenzene

31249-1G 31249-25G 31249-500MG 31249-5G
76887-176EA 126.25 USD
76887-176 76887-178 76887-180 76887-182
2,4-Dichloro-3-fluorobromobenzene ≥97%
2,4-Dichloro-3-fluorobromobenzene
Formula: C₆H₂BrCl₂F
MW: 243.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878169
CAS Number: 1000573-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR