2,4-Dichloro-3-fluorobenzoic acid 95%

Supplier: AOBChem USA

Warning

31245-1G 31245-25G 31245-5G 31245-500MG
76887-164EA 88.06 USD
76887-164 76887-166 76887-168 77645-956
2,4-Dichloro-3-fluorobenzoic acid 95%
2,4-Dichloro-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14707646
CAS Number: 915145-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR