2,3-Dichloro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

31237-1G 31237-25G 31237-500MG 31237-5G
76887-150EA 350.1 USD
76887-150 76887-152 76887-154 76887-156
2,3-Dichloro-4-methylbenzoic acid ≥97%
2,3-Dichloro-4-methylbenzoic acid
Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22628450
CAS Number: 1254073-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR