4-Bromo-2-chloro-5-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

31188-1G 31188-25G 31188-500MG 31188-5G
76887-118EA 189.91 USD
76887-118 76887-120 76887-122 76887-124
4-Bromo-2-chloro-5-fluoroanisole ≥97%
4-Bromo-2-chloro-5-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD16036128
CAS Number: 901236-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR