2-(4-Bromo-2,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

31160-1G 31160-25G 31160-5G
76887-104EA 91.24 USD
76887-104 76887-106 76887-108
2-(4-Bromo-2,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Bromo-2,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₄H₂₀BBrO₂
MW: 311.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22494456
CAS Number: 924964-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR