2-(Trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(2-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane

31122-25G 31122-5G
76887-096EA 304.48 USD
76887-096 76887-098
2-(Trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2-(Trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BF₃O₂
MW: 272.0712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795676
CAS Number: 1073339-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR